missing translation for 'onlineSavingsMsg'
Learn More

Triphenylethylene, 98+%

Product Code. 11430783
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430783 50 g 50g
11420783 10 g 10g
11440783 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430783 Supplier Thermo Scientific Alfa Aesar Supplier No. A13244.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58-72-0 | C20H16 | 256.348 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-72-0
Molecular Formula C20H16
Molecular Weight (g/mol) 256.348
MDL Number MFCD00004765
InChI Key MKYQPGPNVYRMHI-UHFFFAOYSA-N
Synonym triphenylethylene, triphenylethene, ethylene, triphenyl, benzilidenediphenylmethane, 1,1,2-triphenylethylene, ethene-1,1,2-triyltribenzene, 1,2-diphenylvinyl benzene, 1,2,2-triphenylethylene, benzene, 1,1',1-1-ethenyl-2-ylidene tris, unii-s4zlz1k74b
PubChem CID 6025
ChEBI CHEBI:35034
IUPAC Name 1,2-diphenylethenylbenzene
SMILES C1=CC=C(C=C1)C=C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 67°C to 71°C
Boiling Point 219°C to 221°C (14 mmHg)
Quantity 50 g
Beilstein 1867462
Formula Weight 256.35
Percent Purity ≥98%
Chemical Name or Material Triphenylethylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.