missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethylium Tetrafluoroborate, TRC

Product Code. 30456937
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456937 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456937 Supplier Toronto Research Chemicals Supplier No. TRCT8993082.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triphenylmethylium Tetrafluoroborate
CAS 341-02-6
Molecular Formula C19H15BF4
Quantity 2.5 g
Synonym Methylium','triphenyl-','tetrafluoroborate(1-) (1:1) (ACI), Borate(1-)','tetrafluoro-','triphenylmethylium (ZCI), Borate(1-)','tetrafluoro-','tritylium (8CI), Methylium','triphenyl-','tetrafluoroborate(1-) (9CI), Tritylcarbonium fluoborate (6CI), Tritylium tetrafluoroborate (7CI), Tritylium','tetrafluoroborate(1-) (8CI), NSC 176021, Triphenylcarbonium tetrafluoroborate, Triphenylcarbonium tetrafluoroborate(1-), Triphenylmethyl fluoroborate, Triphenylmethyl tetrafluoroborate, Triphenylmethylium tetrafluoroborate, Trityl fluoroborate, Trityl tetrafluoroborate, Trityl tetrafluoroborate(1-)
InChI Formula InChI=1S/C19H15.BF4/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;2-1(3,4)5/h1-15H;/q+1;-1
SMILES F[B-](F)(F)F.c1ccc(cc1)[C+](c2ccccc2)c3ccccc3
IUPAC Name diphenylmethylbenzene;tetrafluoroborate
Molecular Weight (g/mol) 330.1
Formula Weight 330.1203
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.