missing translation for 'onlineSavingsMsg'
Learn More

Triphenylsulfonium chloride, 94%

Product Code. 15403367
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15403367 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15403367 Supplier Thermo Scientific Alfa Aesar Supplier No. H26059.03
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4270-70-6 | C18H15ClS | 298.828 g/mol

Triphenylsulfonium chloride is used as a key intermediate that is widely used in the new electronic chemicals such as semiconductor, LCD and TFT.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylsulfonium chloride is used as a key intermediate that is widely used in the new electronic chemicals such as semiconductor, LCD and TFT.

Solubility
Soluble in water.

Notes
Store in cool pla
TRUSTED_SUSTAINABILITY

Specifications

CAS 4270-70-6
Molecular Formula C18H15ClS
MDL Number MFCD00031716
Quantity 1 g
Synonym triphenylsulfonium chloride, triphenylsulphonium chloride, unii-93vmp8wb2z, sulfonium, triphenyl-, chloride, triphenylsulfanium chloride, triphenyl sulfonium chloride, 93vmp8wb2z, triphenylsulfonium chloride solution, triphenylsulfonium ion, triphenyl sulfide
Solubility Information Soluble in water.
InChI Key ZFEAYIKULRXTAR-UHFFFAOYSA-M
SMILES C1=CC=C(C=C1)[S+](C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
IUPAC Name triphenylsulfanium;chloride
Molecular Weight (g/mol) 298.828
PubChem CID 61343
Formula Weight 298.84
Percent Purity 94%
Chemical Name or Material Triphenylsulfonium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.