missing translation for 'onlineSavingsMsg'
Learn More

Triphenyltin hydride, 97%, Thermo Scientific™

Product Code. 11302685
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302685 5 g 5g
11312685 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302685 Supplier Thermo Scientific Alfa Aesar Supplier No. L00163.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Triphenyltin hydride is used as a source of ″H·″ to generate radicals or cleave carbon-oxygen bonds. And it is also used in anti-fouling paints.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 892-20-6
Molecular Formula C18H16Sn
MDL Number MFCD00003004
Synonym Triphenylstannane

Specifications

Melting Point 26°C to 28°C
Density 1.374
Boiling Point 162°C to 168°C (0.5 mmHg)
Flash Point >110°C (230°F)
Linear Formula (C6H5)3SnH
Refractive Index 1.637
Quantity 5 g
UN Number UN3146
Beilstein 3544353
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 351.02
Percent Purity 97%
Chemical Name or Material Triphenyltin hydride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.