missing translation for 'onlineSavingsMsg'
Learn More

Triphosgene, 98%

Product Code. 11442757
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442757 25 g 25g
11432757 5 g 5g
11452757 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442757 Supplier Thermo Scientific Alfa Aesar Supplier No. A14932.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 32315-10-9 | C3Cl6O3 | 296.73 g/mol

Triphosgene is used as a carbonylating agent for aza-peptide synthesis. It reacts with several alfa-amino acids to give the corresponding N-carboxyanhydrides. It is involved in the preparation of the esterification coupling reagent, di-2-thienyl carbonate from 2(5H)-thiophenone. Further, it is used as a reagent in organic synthesis and converts an amino group into isocyanate. In addition to this, it is employed in the preparation of 2-chloronicotinaldehydes through cyclization of the corresponding enamides. It is considered as a useful substitute for phosgene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphosgene is used as a carbonylating agent for aza-peptide synthesis. It reacts with several alfa-amino acids to give the corresponding N-carboxyanhydrides. It is involved in the preparation of the esterification coupling reagent, di-2-thienyl carbonate from 2(5H)-thiophenone. Further, it is used as a reagent in organic synthesis and converts an amino group into isocyanate. In addition to this, it is employed in the preparation of 2-chloronicotinaldehydes through cyclization of the corresponding enamides. It is considered as a useful substitute for phosgene.

Solubility
Soluble in ether, ethanol, tetrahydrofuran, benzene, cyclohexane, chloroform, hexane, carbon tetrachloride, dichloromethane and chlorobenzene. Insoluble in water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32315-10-9
Molecular Formula C3Cl6O3
Molecular Weight (g/mol) 296.73
MDL Number MFCD00062848
InChI Key UCPYLLCMEDAXFR-UHFFFAOYSA-N
Synonym triphosgene, bis trichloromethyl carbonate, methanol, trichloro-, carbonate 2:1, ditrichloromethyl carbonate, triphosgene bis-trichloromethyl carbonate, methanol, 1,1,1-trichloro-, 1,1'-carbonate, triphosgen, tri phosgene, tri-phosgene
PubChem CID 94429
IUPAC Name bis(trichloromethyl) carbonate
SMILES C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl

Specifications

Melting Point 77°C to 91°C
Density 1.6
Boiling Point 203°C to 206°C
Quantity 25 g
UN Number UN2928
Beilstein 1787583
Sensitivity Moisture sensitive
Solubility Information Soluble in ether,ethanol,tetrahydrofuran,benzene,cyclohexane,chloroform,hexane,carbon tetrachloride,dichloromethane and chlorobenzene. Insoluble in water.
Formula Weight 296.75
Percent Purity 98%
Chemical Name or Material Triphosgene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triphosgene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Triphosgene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.