missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Tripod

Product Code. p-7146779
Click to view available options
Length (Metric):
210 mm
240 mm
Includes:
Screwed Feet
Welded Feet
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10048501

Brand: Bochem™ 5231

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Bochem™ Tripod available with welded feet or screwed feet.

  • Welded feet on Stainless Steel
  • Screwed feet on Tempered Cast Iron

Specifications

Material Tempered Cast Iron
Length (Metric) 210 mm
Diameter (Metric) Inner 120 mm
Includes Screwed Feet
Product Type Tripod
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.