missing translation for 'onlineSavingsMsg'
Learn More

Tripotassium citrate monohydrate, 97%

Product Code. 15402117
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15402117 500 g 500g
15492107 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15402117 Supplier Thermo Scientific Alfa Aesar Supplier No. 045038.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6100-05-6 | C6H7K3O8 | 324.41 g/mol

It is used in the laboratory as a buffering agent and chelator. In renal function studies, potassium citrate is used as an alkanizing agent where the citric acid is a metabolic substrate in renal cells. Studies have shown that potassium citrate may increase bone density, it is therefore used in research experiments related to the treatment of osteoporosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H7K3O8
Molecular Weight (g/mol) 324.41
MDL Number MFCD00150442
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
Synonym potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
PubChem CID 2735208
ChEBI CHEBI:64746
IUPAC Name tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]

Specifications

Melting Point 230°C (decomposition)
Density 1.98 g/mL
Quantity 500 g
Merck Index 14,7623
Solubility Information Soluble in water (640mg/ml at 25°C),and glycerol. Insoluble in alcohol.
Formula Weight 324.42 (306.41 Anhydrous)
Percent Purity 97%
Physical Form Lump or Granular
Chemical Name or Material Tripotassium citrate monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.