missing translation for 'onlineSavingsMsg'
Learn More

Tripotassium Phosphate Monohydrate, TRC

Product Code. 30416201
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30416201 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 30416201

Brand: Toronto Research Chemicals TRCP359488250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tripotassium Phosphate Monohydrate
CAS 27176-10-9
Molecular Formula K3O4P . H2O
Quantity 250 mg
InChI Formula InChI=1S/3K.H3O4P.H2O/c;;;1-5(2,3)4;/h;;;(H3,1,2,3,4);1H2/q3*+1;;/p-3
SMILES O.[K+].[K+].[K+].[O-]P(=O)([O-])[O-]
IUPAC Name tripotassium;phosphate;hydrate
Molecular Weight (g/mol) 230.28
Formula Weight 229.8551
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.