missing translation for 'onlineSavingsMsg'
Learn More

Tripropionin, 97%

Product Code. 10646281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646281 250 g Glass bottle 250g
10551311 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10646281 Supplier Thermo Scientific Acros Supplier No. 275102500
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 139-45-7 | C12H20O6 | 260.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-45-7
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.28
MDL Number MFCD00027005
InChI Key YZWRNSARCRTXDS-UHFFFAOYSA-N
Synonym tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride
PubChem CID 8763
IUPAC Name 2,3-di(propanoyloxy)propyl propanoate
SMILES CCC(=O)OCC(COC(=O)CC)OC(=O)CC

Specifications

Color Colorless
Density 1.0820g/mL
Boiling Point 151°C (2.0 mmHg)
Flash Point 162°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4310 to 1.434
Quantity 250 g
Specific Gravity 1.082
Formula Weight 260.28
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tripropionin, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.