missing translation for 'onlineSavingsMsg'
Learn More

Tripropyl orthoformate, 98%, Thermo Scientific™

Product Code. 10516813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10516813 25 mL Glass bottle 25mL
10622364 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10516813 Supplier Thermo Scientific Acros Supplier No. 369290250
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy
Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-76-1
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.29
MDL Number MFCD00015214
InChI Key RWNXXQFJBALKAX-UHFFFAOYSA-N
Synonym tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate
PubChem CID 69311
IUPAC Name 1-(dipropoxymethoxy)propane
SMILES CCCOC(OCCC)OCCC

Specifications

Color Colorless
Density 0.8790g/mL
Boiling Point 106°C to 108°C (40.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH(OCH2CH2CH3)3
Refractive Index 1.4060 to 1.408
Quantity 25 mL
Beilstein 02, IV, 27
Specific Gravity 0.879
Formula Weight 190.29
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tripropyl orthoformate
Show More Show Less
compliance-icons
Product Identifier
  • Tripropyl orthoformate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.