missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Triptolide

Product Code. 16483746
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483746 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16483746 Supplier MedChemExpress Supplier No. HY32735
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Triptolide is a diterpenoid triepoxide extracted from the root of Tripterygium wilfordii with immunosuppressive, anti-inflammatory, antiproliferative and antitumour effects. Triptolide is a NF-κB activation inhibitor.

Specifications

Content And Storage 4°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
Molecular Weight (g/mol) 360.4 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 38748-32-2
Product Type Natural Products
Purity 98.15%
Solubility DMSO : ≥ 33 mg/mL (91.56 mM) ∣H2O : < 0.1 mg/mL (ultrasonic∣warming∣heat to 60°C) (insoluble)
Synonym PG490
Target NF-κB∣Apoptosis
Quantity 25 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.