missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III)

Product Code. 11359119
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359119 5 g 5g
11345059 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359119 Supplier Thermo Scientific Alfa Aesar Supplier No. 087898.06
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 15522-71-1 | C33H60EuO6 | 704.801 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15522-71-1
Molecular Formula C33H60EuO6
Molecular Weight (g/mol) 704.801
MDL Number MFCD00000019
InChI Key RHXUZKJNHAMZEP-QFVJJVGWSA-N
Synonym eu dpm 3, tris 2,2,6,6-tetramethyl-3,5-heptanedionato europium, tris 2,2,6,6-tetramethyl-3,5-heptanedionato europium iii, resolve-al tm, europium iii-dpm, eu tmhd 3, europium 2,2,6,6-tetramethyl-3,5-heptanedionate, europium-tris dipivaloylmethane, tris 2,2,6,6-tetramethyl-3,5-heptanedionato europium 3
PubChem CID 10897800
IUPAC Name europium;(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Eu]

Specifications

Melting Point 187°C to 189°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 701.78
Physical Form Crystalline
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)europium (III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.