missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)terbium(III), 99%, Thermo Scientific Chemicals

Product Code. 11376249
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11376249 5 g 5g
11366249 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376249 Supplier Thermo Scientific Alfa Aesar Supplier No. 044842.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15492-51-0
Molecular Formula C33H57O6Tb
Molecular Weight (g/mol) 708.74
MDL Number MFCD00064752
InChI Key DIIXZXQKWUIOAN-UHFFFAOYSA-N
Synonym tb tmhd 3, terbium iii tris 2,2,6,6-tetramethyl-3,5-heptanedionate
PubChem CID 133109942
SMILES [Tb+3].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C

Specifications

Melting Point 155°C to 156°C
Quantity 5 g
IUPAC Name terbium(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
Formula Weight 708.74
Percent Purity 99%
Physical Form Granules
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)terbium (III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.