missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III), 99%

Product Code. 11384487
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384487 5 g 5g
11374487 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384487 Supplier Thermo Scientific Alfa Aesar Supplier No. 039581.06
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15492-52-1 | C33H57O6Yb | 722.867 g/mol

It is used as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15492-52-1
Molecular Formula C33H57O6Yb
Molecular Weight (g/mol) 722.867
MDL Number MFCD00000034
InChI Key KACMHCBXMZUMQV-LWTKGLMZSA-K
Synonym 2,2,6,6-tetramethylheptane-3,5-dione; ytterbium 3+, ytterbium 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate, tris 2,2,6,6-tetramethyl-3,5-heptanedionato ytterbium iii-yb reo yb tmhd 3
PubChem CID 44473428
IUPAC Name (Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;ytterbium(3+)
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Yb+3]

Specifications

Melting Point 167°C to 169°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 722.86
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.