missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%

Product Code. 10229203 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10229203 5 g Glass bottle 5g
10154740 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10229203 Supplier Thermo Scientific Acros Supplier No. 208750050
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50525-27-4 | C30H36Cl2N6O6Ru | 748.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50525-27-4
Molecular Formula C30H36Cl2N6O6Ru
Molecular Weight (g/mol) 748.62
MDL Number MFCD00149670
InChI Key WHELTKFSBJNBMQ-UHFFFAOYSA-L
Synonym tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
PubChem CID 131664161
IUPAC Name 2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;hydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1

Specifications

Melting Point >300.0°C
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 748.63
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tris(2, 2'-bipyridyl)ruthenium(II) chloride, hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.