missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-aminoethyl)amine, 98+%

Product Code. 11317251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11317251 500 mL Glass bottle 500mL
10175710 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317251 Supplier Thermo Scientific Supplier No. 314795000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4097-89-6 | C6H18N4 | 146.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4097-89-6
Molecular Formula C6H18N4
Molecular Weight (g/mol) 146.24
InChI Key MBYLVOKEDDQJDY-UHFFFAOYSA-N
Synonym tris 2-aminoethyl amine, tren, taea, 2,2',2-triaminotriethylamine, 1,2-ethanediamine, n,n-bis 2-aminoethyl, tri 2-aminoethyl amine, tren hp, tris aminoethyl amine, nitrilotris ethylamine, n,n-bis 2-aminoethyl ethane-1,2-diamine
PubChem CID 77731
ChEBI CHEBI:30631
IUPAC Name N',N'-bis(2-aminoethyl)ethane-1,2-diamine
SMILES C(CN(CCN)CCN)N

Specifications

Melting Point -16.0°C
Color Yellow
Density 0.9770g/mL
Boiling Point 265.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula (H2NCH2CH2)3N
Refractive Index 1.4960 to 1.4980
Quantity 500 mL
Beilstein 04,II,695
Specific Gravity 0.977
Solubility Information Solubility in water: soluble.
Formula Weight 146.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Tris(2-aminoethyl)amine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tris(2-aminoethyl)amine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.