missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-aminoethyl)amine, 97%

Product Code. 11490437
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490437 500 g 500g
11403900 5 g 5g
11470437 25 g 25g
11480437 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11490437 Supplier Thermo Scientific Alfa Aesar Supplier No. B21789.36
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 4097-89-6 | C6H18N4 | 146.238 g/mol

Tris(2-aminoethyl)amine is a tetradentate chelating ligand and forms stable complexes with transition metals. It is also used as a carbon dioxide absorbent. Further, it acts as a reagent for cleavage of the fluorenylmethyloxycarbonyl (Fmoc) group in peptide synthesis. It reacts with aryl isocyanates and isothiocyanates to give tris-urea and -thiourea derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(2-aminoethyl)amine is a tetradentate chelating ligand and forms stable complexes with transition metals. It is also used as a carbon dioxide absorbent. Further, it acts as a reagent for cleavage of the fluorenylmethyloxycarbonyl (Fmoc) group in peptide synthesis. It reacts with aryl isocyanates and isothiocyanates to give tris-urea and -thiourea derivatives.

Solubility
Miscible with water.

Notes
Air sensitive and hygroscopic. Store in a cool place. Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4097-89-6
Molecular Formula C6H18N4
MDL Number MFCD00008177
InChI Key MBYLVOKEDDQJDY-UHFFFAOYSA-N
PubChem CID 77731
ChEBI CHEBI:30631

Specifications

Density 0.978
pH 11.9
Boiling Point 102°C (0.05 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Linear Formula N(CH2CH2NH2)3
Refractive Index 1.4969
Quantity 500 g
UN Number UN2922
Beilstein 1739626
Sensitivity Air sensitive; Hygroscopic
Synonym tris 2-aminoethyl amine, tren, taea, 2,2',2-triaminotriethylamine, 1,2-ethanediamine, n,n-bis 2-aminoethyl, tri 2-aminoethyl amine, tren hp, tris aminoethyl amine, nitrilotris ethylamine, n,n-bis 2-aminoethyl ethane-1,2-diamine
Solubility Information Miscible with water.
SMILES C(CN(CCN)CCN)N
IUPAC Name N',N'-bis(2-aminoethyl)ethane-1,2-diamine
Molecular Weight (g/mol) 146.238
Formula Weight 146.24
Percent Purity 97%
Chemical Name or Material Tris(2-aminoethyl)amine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tris(2-aminoethyl)amine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: Tris(2-aminoethyl)amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.