missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-aminoethyl)amine, TRC

Product Code. 30455690
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455690 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455690 Supplier Toronto Research Chemicals Supplier No. TRCT8750155G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris(2-aminoethyl)amine
CAS 4097-89-6
Molecular Formula C6 H18 N4
Quantity 5 g
Synonym N1,N1-Bis(2-aminoethyl)-1,2-ethanediamine, 2,2',2''-Nitrilotris[ethylamine], 2,2',2''-Triaminotriethylamine, 4-(2-Aminoethyl)diethylenetriamine, 4-(3-Azapropyl)-1,4,7-triazaheptane, Nitrilotriethylamine, Tren, Tris(β-aminoethyl)amine, β,β',β''-Triaminotriethylamine, N,N,N-Tris(2-aminoethyl)amine, N,N-Bis(2-aminoethyl)-1,2-ethanediamine, N,N-Bis(2-aminoethyl)ethylenediamine
InChI Formula InChI=1S/C6H18N4/c7-1-4-10(5-2-8)6-3-9/h1-9H2
SMILES NCCN(CCN)CCN
IUPAC Name N',N'-bis(2-aminoethyl)ethane-1,2-diamine
Molecular Weight (g/mol) 146.234
Formula Weight 146.153
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.