missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-carboxyethyl)phosphine hydrochloride, 95%, 0.5M soln. in water

Product Code. 15439365
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439365 25 mL 25mL
15429365 5 mL 5mL
15449365 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15439365 Supplier Thermo Scientific Alfa Aesar Supplier No. H51864.AC
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 51805-45-9 | C9H12O6P | 247.16 g/mol

Tris(2-carboxyethyl)phosphine hydrochloride is widely used as a reducing agent to reduce disulfide bonds in proteins and trans-4,5-dihydroxy-1,2-dithiane. It is involved in the reduction of sulfoxides, sulfonyl chlorides, N-oxides and azides. It plays an important role in the tissue homogenization process for RNA isolation. It acts as a chelating agent and forms complexes with various heavy metal ions such as Zn(II), Cd(II), Pb(II), and Ni(II). Furthermore, it is useful in the labeling of cysteine residues with maleimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(2-carboxyethyl)phosphine hydrochloride is widely used as a reducing agent to reduce disulfide bonds in proteins and trans-4,5-dihydroxy-1,2-dithiane. It is involved in the reduction of sulfoxides, sulfonyl chlorides, N-oxides and azides. It plays an important role in the tissue homogenization process for RNA isolation. It acts as a chelating agent and forms complexes with various heavy metal ions such as Zn(II), Cd(II), Pb(II), and Ni(II). Furthermore, it is useful in the labeling of cysteine residues with maleimides.

Solubility
Miscible with water and methanol.

Notes
Air sensitive. Store in cool place. Incompatible with strong oxidizing agents
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 51805-45-9
Molecular Formula C9H12O6P
Molecular Weight (g/mol) 247.16
MDL Number MFCD00145469
InChI Key PZBFGYYEXUXCOF-UHFFFAOYSA-K
Synonym tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
PubChem CID 2734570
IUPAC Name 3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
SMILES [O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O

Especificaciones

Linear Formula (HO2CCH2CH2)3P·HCl
Quantity 25 mL
UN Number UN3265
Solubility Information Miscible with water and methanol.
Formula Weight 286.65
Percent Purity 95%
Concentration or Composition (by Analyte or Components) 0.5M soln. in water
Chemical Name or Material Tris(2-carboxyethyl)phosphine hydrochloride
compliance-icons
Nombre del producto
  • Tris(2-carboxyethyl)phosphine hydrochloride 0.5M aqueous solution
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Otros peligros
  • MIXTURE LIST-Contain: Tris(2-carboxyethyl)phosphonium chloride

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.