missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade

Product Code. 15460127
Change view
Click to view available options
Quantity:
2 g
5 g
10 g
Unit Size:
10g
2g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460127 5 g 5g
15450127 2 g 2g
15470127 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15460127 Supplier Thermo Scientific Chemicals Supplier No. J60316.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51805-45-9 | C9H12O6P | 247.16 g/mol

For reducing disulfide bonds in peptides and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51805-45-9
Molecular Formula C9H12O6P
Molecular Weight (g/mol) 247.16
MDL Number MFCD00145469
InChI Key PZBFGYYEXUXCOF-UHFFFAOYSA-K
Synonym tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
PubChem CID 2734570
IUPAC Name 3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
SMILES [O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O

Specifications

Melting Point 177°C to 178°C
Packaging Plastic bottle
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Soluble in DMSO or water
Formula Weight 286.65
Percent Purity 98%
Grade Molecular Biology
Physical Form Crystalline Powder
Chemical Name or Material Tris(2-carboxyethyl)phosphine hydrochloride
Show More Show Less
compliance-icons
Identificativo prodotto
  • Tris(2-carboxyethyl)phosphine hydrochloride, Molecular Biology Grade
Avvertenza
  • Danger
Categoria di pericolo
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Indicazioni di pericolo
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Informazioni supplementari
  • MIXTURE LIST-Contain: Tris(2-carboxyethyl)phosphonium chloride

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.