missing translation for 'onlineSavingsMsg'
Learn More

Tris (2-cyanoethyl)phosphine, 95%

Product Code. 15274774 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15274774 50 g 50g
15254774 1 g 1g
15264774 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15274774 Supplier Thermo Scientific Acros Supplier No. 449710500
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4023-53-4 | C9H12N3P | 193.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4023-53-4
Molecular Formula C9H12N3P
Molecular Weight (g/mol) 193.19
InChI Key CHZAMJVESILJGH-UHFFFAOYSA-N
Synonym tris 2-cyanoethyl phosphine, 3,3',3-phosphinetriyltripropanenitrile, tris-2-cyanoethyl phosphine, tris-2-cyanoethyl-phosphine, propanenitrile, 3,3',3-phosphinidynetris, acmc-20ajc3, p ch2ch2cn 3, 3-bis 2-cyanoethyl phosphanyl propanenitrile, propanenitrile,3',3-phosphinidynetris, tris 2-cyanoethyl phosphine 10g
PubChem CID 77639
IUPAC Name 3-[bis(2-cyanoethyl)phosphanyl]propanenitrile
SMILES C(CP(CCC#N)CCC#N)C#N

Specifications

Melting Point 97.0°C to 99.0°C
Boiling Point 235.0°C (0.9 mmHg)
Assay Percent Range 94% min. (Iodimetry)
Packaging Glass bottle
Quantity 50 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 193.19
Percent Purity 95%
Chemical Name or Material Tris (2-cyanoethyl)phosphine
compliance-icons compliance-icons
Product Identifier
  • Tris (2-cyanoethyl)phosphine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
  • P313-Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.