missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-dimethylaminoethyl)amine, 98+%

Product Code. 15424507
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15424507 5 g 5g
15434507 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424507 Supplier Thermo Scientific Alfa Aesar Supplier No. H31853.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33527-91-2 | C12H30N4 | 230.40 g/mol

Tris(2-dimethylaminoethyl)amine is used as an atom transfer radical polymerization (ATRP) ligand for the creation of telechelic polymers. It is a tripodal polyamine ligand and forms complexes with group 1 metals, which is used in alkali-metal-mediated synthetic applications. Further, it is involved in the macrocyclization processes by reacting with tripodal esters in methanol to give tricyclic cryptands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(2-dimethylaminoethyl)amine is used as an atom transfer radical polymerization (ATRP) ligand for the creation of telechelic polymers. It is a tripodal polyamine ligand and forms complexes with group 1 metals, which is used in alkali-metal-mediated synthetic applications. Further, it is involved in the macrocyclization processes by reacting with tripodal esters in methanol to give tricyclic cryptands.

Solubility
Miscible with water.

Notes
Hygroscopic. Air sensitive. Incompatible with oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33527-91-2
Molecular Formula C12H30N4
Molecular Weight (g/mol) 230.40
MDL Number MFCD00015607
InChI Key VMGSQCIDWAUGLQ-UHFFFAOYSA-N
Synonym n1,n1-bis 2-dimethylamino ethyl-n2,n2-dimethylethane-1,2-diamine, tris 2-dimethylaminoethyl amine, tris 2-dimethylamino ethyl amine, me6tren, 1,2-ethanediamine, n,n-bis 2-dimethylamino ethyl-n',n'-dimethyl, 2-bis 2-dimethylamino ethyl amino ethyl dimethylamine, acmc-20ah4b, n,n-bis 2-dimethylamino ethyl-n',n'-dimethylethane-1,2-diamine
PubChem CID 263094
IUPAC Name N',N'-bis[2-(dimethylamino)ethyl]-N,N-dimethylethane-1,2-diamine
SMILES CN(C)CCN(CCN(C)C)CCN(C)C

Specifications

Boiling Point 70°C to 71°C (0.5 mmHg)
Linear Formula [(CH3)2NCH2CH2]3N
Refractive Index 1.454
Quantity 5 g
UN Number UN2735
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Formula Weight 230.4
Percent Purity ≥99%
Chemical Name or Material Tris(2-dimethylaminoethyl)amine
compliance-icons
Product Identifier
  • Tris(2-dimethylaminoethyl)amine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Tris(2-dimethylaminoethyl)amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.