missing translation for 'onlineSavingsMsg'
Learn More

Tris[2-(diphenylphosphino)ethyl]phosphine, 97%, Thermo Scientific Chemicals

Catalog Number 10139931
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
23582-03-8
C42H42P4
670.69
MFCD00010009
TVLNGWSWPKIYAO-UHFFFAOYSA-N
tris 2-diphenylphosphino ethyl phosphine, tris 2-diphenylphosphinoethyl phosphine, tetraphos-li, 2-bis 2-diphenylphosphanyl ethyl phosphanyl ethyl diphenylphosphane, acmc-20ajaw, pubchem6541, chembl65725, tris 2-diphenylphosphanylethyl phosphane
98777
tris(2-diphenylphosphanylethyl)phosphane
C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
23582-03-8
C42H42P4
670.69
MFCD00010009
TVLNGWSWPKIYAO-UHFFFAOYSA-N
tris 2-diphenylphosphino ethyl phosphine, tris 2-diphenylphosphinoethyl phosphine, tetraphos-li, 2-bis 2-diphenylphosphanyl ethyl phosphanyl ethyl diphenylphosphane, acmc-20ajaw, pubchem6541, chembl65725, tris 2-diphenylphosphanylethyl phosphane
98777
tris(2-diphenylphosphanylethyl)phosphane
C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 134.0°C to 139.0°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula [(C6H5)2PCH2CH2]3P
Quantity 1 g
Formula Weight 670.69
Percent Purity 97%
Chemical Name or Material Tris[2-(diphenylphosphino)ethyl]phosphine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'