missing translation for 'onlineSavingsMsg'
Learn More

Tris[2-(diphenylphosphino)ethyl]phosphine, 97%

Product Code. 11975841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11975841 25 g 25g
10139931 1 g 1g
10147463 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11975841 Supplier Thermo Scientific Acros Supplier No. 316830250
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23582-03-8 | C42H42P4 | 670.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23582-03-8
Molecular Formula C42H42P4
Molecular Weight (g/mol) 670.69
MDL Number MFCD00010009
InChI Key TVLNGWSWPKIYAO-UHFFFAOYSA-N
Synonym tris 2-diphenylphosphino ethyl phosphine, tris 2-diphenylphosphinoethyl phosphine, tetraphos-li, 2-bis 2-diphenylphosphanyl ethyl phosphanyl ethyl diphenylphosphane, acmc-20ajaw, pubchem6541, chembl65725, tris 2-diphenylphosphanylethyl phosphane
PubChem CID 98777
IUPAC Name tris(2-diphenylphosphanylethyl)phosphane
SMILES C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 134.0°C to 139.0°C
Assay Percent Range 97%
Linear Formula [(C6H5)2PCH2CH2]3P
Quantity 25 g
Formula Weight 670.69
Percent Purity 97%
Chemical Name or Material Tris[2-(diphenylphosphino)ethyl]phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.