missing translation for 'onlineSavingsMsg'
Learn More

tris(2-Ethylhexyl)phosphate, 99.0%, MP Biomedicals™

Product Code. 11435431 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435431 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11435431 Supplier MP Biomedicals Supplier No. 0215799125
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 78-42-2
Color Colorless
Density 0.926 at 20°C
Assay Percent Range 99%
Molecular Formula C24H51O4P
MDL Number MFCD00009491
Quantity 25 mL
Synonym tris 2-ethylhexyl phosphate, disflamoll tof, flexol tof, phosphoric acid, tris 2-ethylhexyl ester, kronitex tof, flexol plasticizer tof, 2-ethyl-1-hexanol phosphate, tris ethylhexyl phosphate, tri 2-ethylhexyl phosphate
InChI Key GTVWRXDRKAHEAD-UHFFFAOYNA-N
SMILES CCCCC(CC)COP(=O)(OCC(CC)CCCC)OCC(CC)CCCC
IUPAC Name tris(2-ethylhexyl) phosphate
Molecular Weight (g/mol) 434.64
PubChem CID 6537
Formula Weight 434.63
Physical Form Liquid
Chemical Name or Material tris(2-Ethylhexyl)phosphate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.