missing translation for 'onlineSavingsMsg'
Learn More

Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride

Product Code. 11319608
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11319608 2 g 2g
11309608 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319608 Supplier Thermo Scientific Alfa Aesar Supplier No. 044123.04
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Redox indicator, oxygen sensor | CAS: 36309-88-3 | C72H48Cl2N6Ru | 1169.188 g/mol

Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride is used as a probe for luminescent detection and quantitation of oxygen. It is also used for fiber optic sensors. Further, it is used in studies on oxygen in skin and skin tumors as well as measurement of oxygen flux through the skin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36309-88-3
Molecular Formula C72H48Cl2N6Ru
Molecular Weight (g/mol) 1169.188
MDL Number MFCD03095387
InChI Key SKZWFYFFTOHWQP-UHFFFAOYSA-L
Synonym bathophenanthroline ruthenium chloride, ru ddp, tris-bathophenanthroline ruthenium ii chloride, dichlororuthenium-4,7-diphenyl-1,10-phenanthroline 1/3, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride, tris 4,7-diphenyl-1,10-phenanthroline ; dichlororuthenium, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, for fluorescence
PubChem CID 15787787
IUPAC Name dichlororuthenium;4,7-diphenyl-1,10-phenanthroline
SMILES C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.Cl[Ru]Cl

Specifications

Quantity 2 g
Solubility Information Soluble in alcohol and dimethylformamide. Insoluble in water.
Formula Weight 1169.2
Physical Form Powder
Chemical Name or Material Tris (4,7-diphenyl-1,10-phenanthroline)ruthenium (II) dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.