missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-trifluoromethylphenyl)phosphine, TRC

Product Code. 30426200
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426200 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426200 Supplier Toronto Research Chemicals Supplier No. TRCT245170250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Tris(4-trifluoromethylphenyl)phosphine
CAS 13406-29-6
Molecular Formula C21 H12 F9 P
Quantity 250 mg
Synonym Phosphine','tris(alpha,alpha,alpha-trifluoro-p-tolyl)- (7CI,8CI), Tris[p-(trifluoromethyl)phenyl]phosphine
InChI Formula InChI=1S/C21H12F9P/c22-19(23,24)13-1-7-16(8-2-13)31(17-9-3-14(4-10-17)20(25,26)27)18-11-5-15(6-12-18)21(28,29)30/h1-12H
SMILES FC(F)(F)c1ccc(cc1)P(c2ccc(cc2)C(F)(F)F)c3ccc(cc3)C(F)(F)F
IUPAC Name tris[4-(trifluoromethyl)phenyl]phosphane
Molecular Weight (g/mol) 466.279
Formula Weight 466.053
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.