missing translation for 'onlineSavingsMsg'
Learn More

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)ytterbium(III), 99% (metals basis)

Product Code. 11304497
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304497 1 g 1g
11394487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304497 Supplier Thermo Scientific Alfa Aesar Supplier No. 039582.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 18323-96-1 | C30H30F21O6Yb | 1058.584 g/mol

It is a reactant involved in the synthesis of rare earth metal complexes for separation of lanthanide metals, conformational mobility of substituted 2-methoxychalcones under the action of lanthanide shift reagents, NMR titration on cation-π complexes of bowl-shaped polycyclic aromatic hydrocarbons, specific recognition of chiral amino alcohols. It finds its application as catalysis and synthesis, optics & glasses, healthcare & biochemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18323-96-1
Molecular Formula C30H30F21O6Yb
Molecular Weight (g/mol) 1058.584
MDL Number MFCD00044289
InChI Key KZBQCXBCJMHJOB-VNGPFPIXSA-K
Synonym tris 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato ytterbium, 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyloctane-3,5-dione; ytterbium 3+, z-6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-5-oxooct-3-en-3-olate; ytterbium 3+
PubChem CID 6510275
IUPAC Name (Z)-6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-5-oxooct-3-en-3-olate;ytterbium(3+)
SMILES CC(C)(C)C(=CC(=O)C(C(C(F)(F)F)(F)F)(F)F)[O-].CC(C)(C)C(=CC(=O)C(C(C(F)(F)F)(F)F)(F)F)[O-].CC(C)(C)C(=CC(=O)C(C(C(F)(F)F)(F)F)(F)F)[O-].[Yb+3]

Specifications

Melting Point 108°C to 111°C
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Slowly degrades in water and soluble in hydrocarbons,alcohols,ketones,esters.
Formula Weight 1058.59
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)ytterbium(III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.