missing translation for 'onlineSavingsMsg'
Learn More

Tris(cyclopentadienyl)holmium(III), 98%

Product Code. 11418400
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418400 5 g 5g
11408400 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418400 Supplier Thermo Scientific Alfa Aesar Supplier No. 044302.06
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1272-22-6 | C15H15Ho | 360.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1272-22-6
Molecular Formula C15H15Ho
Molecular Weight (g/mol) 360.215
MDL Number MFCD03427140
InChI Key ZZSQGMHUOVLCBI-UHFFFAOYSA-N
Synonym tris cyclopentadienyl holmium, acmc-1bt45, holmium,tris h5-2,4-cyclopentadien-1-yl, tris, c5-cyclopenta-2,4-dien-1-yl holmium, tris cyclopenta-1,3-dien-1-yl holmium
PubChem CID 21115617
IUPAC Name cyclopenta-1,3-diene;holmium(3+)
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Ho+3]

Specifications

Melting Point 295°C
Boiling Point 230°C (sublimation)(vac.)
Packaging Ampule
Quantity 5 g
UN Number UN3181
Sensitivity Moisture sensitive
Formula Weight 360.21
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Tris (cyclopentadienyl)holmium (III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.