missing translation for 'onlineSavingsMsg'
Learn More

Tris(cyclopentadienyl)yttrium(III), 99.9% (REO)

Product Code. 11342428
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
This item is not returnable. View return policy

Product Code. 11342428

Brand: Thermo Scientific Alfa Aesar 014478.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1294-07-1 | C15H15Y | 284.191 g/mol

Tris(cyclopentadienyl)yttrium(III) is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1294-07-1
Molecular Formula C15H15Y
Molecular Weight (g/mol) 284.191
MDL Number MFCD00058706
InChI Key IUJOPJIXAVSZIQ-UHFFFAOYSA-N
Synonym yttrium 3+ ion tris cyclopenta-1,3-dien-1-ide
PubChem CID 22481132
IUPAC Name cyclopenta-1,3-diene;yttrium(3+)
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Y+3]

Specifications

Melting Point 295°C
Boiling Point 200°C (sublimation)(vacuum)
Packaging Ampule
Quantity 2 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 284.2
Percent Purity 99.9%
Physical Form Powder
Assay (REO)
Chemical Name or Material Tris(cyclopentadienyl)yttrium(III)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.