missing translation for 'onlineSavingsMsg'
Learn More

Tris(ethylenediamine)cobalt chloride trihydrate, 97%, Thermo Scientific™

Product Code. 15510604 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15510604

Brand: Thermo Scientific Acros 458960050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 14883-80-8
Molecular Formula C6H24Cl3CoN6·3H2O
Molecular Weight (g/mol) 399.64
InChI Key WBMVTRUQSMZIKW-UHFFFAOYSA-K
Synonym tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride
PubChem CID 73995044
IUPAC Name cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate
SMILES C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3]

Specifications

Melting Point >300.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 399.64
Percent Purity 97%
Chemical Name or Material Tris(ethylenediamine)cobalt chloride trihydrate
compliance-icons
Product Identifier
  • Tris(ethylenediamine)cobalt chloride trihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.