missing translation for 'onlineSavingsMsg'
Learn More

Tris(ethylenediamine)rhodium(III) chloride trihydrate, Rh 23.2%

Product Code. 11336068
Click to view available options
Quantity:
0.5 g
1 g
5 g
Unit Size:
0.5g
1g
5g
This item is not returnable. View return policy

Product Code. 11336068

Brand: Thermo Scientific Alfa Aesar 010553.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15004-86-1 | C6H30Cl3N6O3Rh | 443.60 g/mol

Used as precursor for supported RH catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15004-86-1
Molecular Formula C6H30Cl3N6O3Rh
Molecular Weight (g/mol) 443.60
MDL Number MFCD00149661
InChI Key HHSLUQMEKOPKJG-UHFFFAOYSA-K
Synonym Trichlorotris(ethylenediamine)rhodium(III)
IUPAC Name rhodium(3+) tris(ethane-1,2-diamine) trihydrate trichloride
SMILES O.O.O.[Cl-].[Cl-].[Cl-].[Rh+3].NCCN.NCCN.NCCN

Specifications

Melting Point 280°C (decomposition)
Color White
Linear Formula Rh(H2NCH2CH2NH2)3Cl3·3H2O
Quantity 1 g
Formula Weight 443.61 (389.56 Anhydrous)
Concentration or Composition (by Analyte or Components) Rh 23.2%
Physical Form Crystalline/Fused Lump, Crystals Fuse Over Time
Chemical Name or Material Tris(ethylenediamine)rhodium(III) chloride trihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.