missing translation for 'onlineSavingsMsg'
Learn More

Tris(hydroxymethyl)aminomethane, 99%

Product Code. 11448407
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448407 100 g 100g
11458407 500 g 500g
11468407 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448407 Supplier Thermo Scientific Alfa Aesar Supplier No. A18494.22
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

Tris(hydroxymethyl)aminomethane is used as buffers in biochemistry and molecular biology laboratories. It is used as a primary standard to standardize acid solutions for chemical analysis. It finds application in cell membranes to increase its permeability. As an alternative to sodium bicarbonate, it used in the treatment of metabolic acidosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(hydroxymethyl)aminomethane is used as buffers in biochemistry and molecular biology laboratories. It is used as a primary standard to standardize acid solutions for chemical analysis. It finds application in cell membranes to increase its permeability. As an alternative to sodium bicarbonate, it used in the treatment of metabolic acidosis.

Solubility
Soluble in water, ethylene glycol, methanol, ethanol,dimethyl formamide, acetone, ethyl acetate, olive oil, chloroform and alkaline solution.

Notes
Hygroscopic. Incompatible with bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 168°C to 172°C
Density 1.353
Boiling Point 219°C to 220°C (10 mmHg)
Linear Formula (HOCH2)3CNH2
Quantity 100 g
Beilstein 741883
Sensitivity Hygroscopic
Merck Index 14,9772
Solubility Information Soluble in water,ethylene glycol,methanol,ethanol,dimethyl formamide,acetone,ethyl acetate,olive oil,chloroform and alkaline solution.
Formula Weight 121.14
Percent Purity 99%
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.