missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane hydrochloride, 99+%

Product Code. 11420203
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420203 250 g 250g
11410203 50 g 50g
11400203 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420203 Supplier Thermo Scientific Chemicals Supplier No. A11379.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00012590
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Melting Point ∼152°C (decomposition)
pH 3.5 to 5.0
Linear Formula (HOCH2)3CNH2·HCl
Quantity 250 g
Beilstein 3675235
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 157.6
Percent Purity ≥99%
Chemical Name or Material Tris (hydroxymethyl)aminomethane hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.