missing translation for 'onlineSavingsMsg'
Learn More

Tris(hydroxymethyl)nitromethane, 95%

Product Code. 11328066
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328066 500 g 500g
11318066 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328066 Supplier Thermo Scientific Alfa Aesar Supplier No. L13349.36
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 126-11-4 | C4H9NO5 | 151.12 g/mol

Tris(hydroxymethyl)nitromethane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(hydroxymethyl)nitromethane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a primary and secondary intermediate.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-11-4
Molecular Formula C4H9NO5
Molecular Weight (g/mol) 151.12
MDL Number MFCD00007395
InChI Key OLQJQHSAWMFDJE-UHFFFAOYSA-N
Synonym tris hydroxymethyl nitromethane, 2-hydroxymethyl-2-nitropropane-1,3-diol, 2-hydroxymethyl-2-nitro-1,3-propanediol, cimcool wafers, tris nitro, trimethylolnitromethane, isobutylglycerol, nitro, nitroisobutylglycerol, 1,3-propanediol, 2-hydroxymethyl-2-nitro, methane, trimethylolnitro
PubChem CID 31337
IUPAC Name 2-(hydroxymethyl)-2-nitropropane-1,3-diol
SMILES OCC(CO)(CO)[N+]([O-])=O

Specifications

Melting Point ∽160°C (decomposition)
Linear Formula (HOCH2)3CNO2
Quantity 500 g
Beilstein 742427
Merck Index 14,9753
Solubility Information It is soluble in water.
Formula Weight 151.12
Percent Purity 95%
Chemical Name or Material Tris(hydroxymethyl)nitromethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.