missing translation for 'onlineSavingsMsg'
Learn More

Tris[N,N-bis(trimethylsilyl)amide]cerium(III), 98%, Thermo Scientific Chemicals

Product Code. 11478520
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478520 5 g 5g
11400710 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478520 Supplier Thermo Scientific Alfa Aesar Supplier No. 044362.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 41836-21-9 | Molecular Formula: C18H54CeN3Si6 | Molecular Weight: 621.277 g/mol

Tris[N,N-bis(trimethylsilyl)amide]cerium(III) is used as a reactant for synthesis of amidocerium(IV) oxides, Sterically encumbered N-heterocyclic carbene ligands, Lanthanide imidodiphosphinate complexes with electroluminescent properties, Lanthanide complexes of an oxazoline-phenoxide hybrid chelate. Some of the other applications include it is used as a reactant for One-electron oxidation to produce tetravalent cerium compounds, Addition-elimination reactions across the M-C bond of metal N-heterocyclic carbenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41836-21-9
Molecular Formula C18H54CeN3Si6
Molecular Weight (g/mol) 621.277
MDL Number MFCD03411241
InChI Key DPCZDYUESIKMSU-UHFFFAOYSA-N
Synonym bis trimethylsilyl azanide; cerium 3+, tris bis trimethylsilyl amino cerium iii, cerium 3+ tris bis trimethylsilyl azanide, tris n,n-bis trimethylsilyl amide cerium iii , 96 %
PubChem CID 6093781
IUPAC Name bis(trimethylsilyl)azanide;cerium(3+)
SMILES C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Ce+3]

Specifications

Color Yellow
Linear Formula [[(CH3)3Si]2N]3Ce
Quantity 5 g
UN Number UN3396
Sensitivity Moisture Sensitive
Formula Weight 621.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]cerium (III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.