missing translation for 'onlineSavingsMsg'
Learn More

Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III), 97%

Product Code. 11468560
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468560 1 g 1g
11478560 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468560 Supplier Thermo Scientific Alfa Aesar Supplier No. 044381.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 35788-99-9 | C18H54LaN3Si6 | 620.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35788-99-9
Molecular Formula C18H54LaN3Si6
Molecular Weight (g/mol) 620.066
MDL Number MFCD00271009
InChI Key ZDYNTRMQDURVDM-UHFFFAOYSA-N
Synonym lanthanum tris bis trimethylsilyl amide, tris bis trimethylsilyl amino lanthanum, bis trimethylsilyl azanide; lanthanum 3+, acmc-20ako2, lanthanum tris hexamethyldisilazide, lanthanum 3+ tris bis trimethylsilyl azanide, lanthanum tris trimethyl-n-trimethylsilyl silanaminide, lanthanum iii tris n,n-bis trimethylsiyl amide, silanamine,1,1,1-trimethyl-n-trimethylsilyl-, lanthanum 3+ salt 3:1
PubChem CID 6093756
IUPAC Name bis(trimethylsilyl)azanide;lanthanum(3+)
SMILES C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[La+3]

Specifications

Melting Point 149°C to 152°C
Color White
Flash Point 2°C (36°F)
Linear Formula [[(CH3)3Si]2N]3La
Quantity 1 g
UN Number UN3396
Sensitivity Moisture sensitive
Formula Weight 620.06
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]lanthanum (III)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.