missing translation for 'onlineSavingsMsg'
Learn More

Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III), 97%

Product Code. 11478560
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478560 5 g 5g
11468560 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478560 Supplier Thermo Scientific Alfa Aesar Supplier No. 044381.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 35788-99-9 | C18H54LaN3Si6 | 620.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35788-99-9
Molecular Formula C18H54LaN3Si6
Molecular Weight (g/mol) 620.066
MDL Number MFCD00271009
InChI Key ZDYNTRMQDURVDM-UHFFFAOYSA-N
Synonym lanthanum tris bis trimethylsilyl amide, tris bis trimethylsilyl amino lanthanum, bis trimethylsilyl azanide; lanthanum 3+, acmc-20ako2, lanthanum tris hexamethyldisilazide, lanthanum 3+ tris bis trimethylsilyl azanide, lanthanum tris trimethyl-n-trimethylsilyl silanaminide, lanthanum iii tris n,n-bis trimethylsiyl amide, silanamine,1,1,1-trimethyl-n-trimethylsilyl-, lanthanum 3+ salt 3:1
PubChem CID 6093756
IUPAC Name bis(trimethylsilyl)azanide;lanthanum(3+)
SMILES C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[La+3]

Specifications

Melting Point 149°C to 152°C
Color White
Flash Point 2°C (36°F)
Linear Formula [[(CH3)3Si]2N]3La
Quantity 5 g
UN Number UN3396
Sensitivity Moisture sensitive
Formula Weight 620.06
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]lanthanum (III)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H228-Flammable solid.
  • H261-In contact with water releases flammable gases.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
Supplemental information
  • MIXTURE LIST-Contain: Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.