missing translation for 'onlineSavingsMsg'
Learn More

Tris-phosphate, 98%

Product Code. 10185060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10185060 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10185060 Supplier Thermo Scientific Acros Supplier No. 264952500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6992-39-8 | C4H14NO7P | 219.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6992-39-8
Melting Point 168.0°C to 172.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C4H14NO7P
Linear Formula NH2C(CH2OH)3·H3PO4
MDL Number MFCD00043285
Quantity 250 g
Synonym tris phosphate, tris phosphate monobasic, tris hydroxymethyl aminomethane phosphate, 2-hydroxy-1,1-bis hydroxymethyl ethyl ammonium dihydrogen phosphate, 2-amino-2-hydroxymethyl propane-1,3-diol; phosphoric acid, mono tris hydroxymethyl aminomethane phosphate, phosphoric acid; tris buffer, trisphosphate, c4h11no3.h3o4p, mono-tris phosphate
Solubility Information Solubility in water: 1g/100 mL.
InChI Key JLEXUIVKURIPFI-UHFFFAOYSA-N
SMILES OP(O)(O)=O.NC(CO)(CO)CO
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;phosphoric acid
Molecular Weight (g/mol) 219.13
PubChem CID 3083992
ChEBI CHEBI:67098
Formula Weight 219.13
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tris-phosphate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.