missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris SDS PAGE Running Buffer, FASTRun, 10X
Description
- Cut the time you spend waiting for your SDS PAGE gels to finish by over 70% while providing comparable or better clarity over traditional buffers
- Novel buffer chemistry gives you better resolution, thus requiring fewer TG gel compositions to fully resolve a protein.
Specifications
Specifications
| CAS | 151-21-3 |
| Color | Colorless to Yellow |
| pH Range | 7.7 to 8.3 (1X/10X soln. at 25°C) |
| Form | Liquid |
| Purity or Quality Grade | Protein Electrophoresis |
| Quantity | 500mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?