missing translation for 'onlineSavingsMsg'
Learn More

Tris(trifluoromethylhydroxymethylene-d-camphorato)europium(III)

Product Code. 11339279
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11339279 5 g 5g
11329279 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11339279

Brand: Thermo Scientific Alfa Aesar 088481.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34830-11-0 | C36H45EuF9O6 | 896.7 g/mol

Optically active NMR shift reagent. Used with Ag(fod) for the enantiomeric resolution of sec-butylisothiouronium chloride. Also reactant involved in synthesis of europium acetyl camphorate phosphine oxide complexes and reactions with 1,10-N,N-phenanthroline dioxide derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34830-11-0
Molecular Formula C36H45EuF9O6
Molecular Weight (g/mol) 896.7
MDL Number MFCD00074803
InChI Key YXJPWWGLCOPUGI-UFRADDTFSA-N
Synonym eu tfc 3, eu facam 3, europium tris 3-trifluoromethylhydroxymethylene---camphorate, europium tris 3-trifluoromethylhydroxymethylene-+-camphorate, tris 3-trifluoromethylhydroxymethylene-d-camphorato europium iii, tris 3-trifluoroacetyl-d-camphorato europium iii, europium iii tris 3-trifluoromethylhydroxymethylene-d-camphorate
PubChem CID 71310195
IUPAC Name europium;(2E)-4,7,7-trimethyl-2-(2,2,2-trifluoro-1-hydroxyethylidene)bicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.[Eu]

Specifications

Melting Point 195°C to 198°C
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform. Insoluble in water.
Formula Weight 893.68
Physical Form Crystalline
Chemical Name or Material Tris (trifluoromethylhydroxymethylene-d-camphorato)europium (III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.