missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl) borate, 98%

Product Code. 11332446
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332446 25 g 25g
11322446 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332446 Supplier Thermo Scientific Alfa Aesar Supplier No. L06277.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4325-85-3 | C9H27BO3Si3 | 278.377 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4325-85-3
Molecular Formula C9H27BO3Si3
Molecular Weight (g/mol) 278.377
MDL Number MFCD00051588
InChI Key YZYKZHPNRDIPFA-UHFFFAOYSA-N
Synonym tris trimethylsilyl borate, tris trimethylsiloxy boron, silanol, trimethyl-, triester with boric acid h3bo3, tritrimethylsilyl borate, borane, tris trimethylsiloxy, silanol, 1,1,1-trimethyl-, 1,1',1-triester with boric acid h3bo3, acmc-209jtk, tris trimethylsiloxy borane, boric acid, 3tms derivative
PubChem CID 78020
IUPAC Name tris(trimethylsilyl) borate
SMILES B(O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Melting Point -35°C
Density 0.834
Boiling Point 183°C to 185°C
Flash Point 42°C (107°F)
Refractive Index 1.3861
Quantity 25 g
UN Number UN1993
Beilstein 1772733
Sensitivity Moisture sensitive
Formula Weight 278.38
Percent Purity 98%
Chemical Name or Material Tris(trimethylsilyl) borate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.