missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl) phosphite, 95%

Product Code. 11353327
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353327 0.5 g 0.5g
11363327 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353327 Supplier Thermo Scientific Alfa Aesar Supplier No. 017159.02
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1795-31-9 | C9H27O3PSi3 | 298.541 g/mol

Tris(trimethylsilyl) phosphite is a reagent used for the mild preparation of bis(TMS) phosphonates (Arbuzow reaction). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1795-31-9
Molecular Formula C9H27O3PSi3
Molecular Weight (g/mol) 298.541
MDL Number MFCD00015588
InChI Key VMZOBROUFBEGAR-UHFFFAOYSA-N
Synonym tris trimethylsilyl phosphite, phosphorous acid tris trimethylsilyl ester, silanol, trimethyl-, phosphite 3:1, tritrimethylsilyl phosphite, tmspi, acmc-209efr, trimethylsilanol phosphite, tris-trimethylsilylphosphite, tris trimethylsily phosphite
PubChem CID 137213
IUPAC Name tris(trimethylsilyl) phosphite
SMILES C[Si](C)(C)OP(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Density 0.893 g/mL
Boiling Point 80°C (8 mmHg)
Refractive Index 1.409
Quantity 0.5 g
Sensitivity Air and moisture sensitive
Solubility Information Sparingly soluble in water.
Formula Weight 298.54
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyl) phosphite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.