missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl)silane, 97%

Product Code. 11410627
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11410627

Brand: Thermo Scientific Alfa Aesar B22457.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1873-77-4 | C9H28Si4 | 248.66 g/mol

Tris(trimethylsilyl)silane is used in hydrosilylations of carbonyls, radical reactions, reductions of acid chlorides, and carbon-halogen bonds. It acts a mediator of radical reactions and considered to be a nontoxic substitute for tri-n-butylstannane in radical reactions. It forms complexes with transition metals and main group elements. Further, it is utilized in carbon-carbon bond forming reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(trimethylsilyl)silane is used in hydrosilylations of carbonyls, radical reactions, reductions of acid chlorides, and carbon-halogen bonds. It acts a mediator of radical reactions and considered to be a nontoxic substitute for tri-n-butylstannane in radical reactions. It forms complexes with transition metals and main group elements. Further, it is utilized in carbon-carbon bond forming reactions.

Solubility
Miscible with pentane, ether, toluene and tetrahydrofuran. Sparingly miscible with acetone and acetonitrile. Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1873-77-4
Molecular Formula C9H28Si4
Molecular Weight (g/mol) 248.66
MDL Number MFCD00077893
InChI Key SQMFULTZZQBFBM-UHFFFAOYSA-N
Synonym tris trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane, tris trimethylsilyl silicon, ttmss, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl-trisilane, hypersilyl, tris trimethylsilyl silyl radical, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane #, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilan-2-yl
IUPAC Name 1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
SMILES C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C

Specifications

Boiling Point 82°C to 84°C (12mmHg)
Quantity 5 g
UN Number UN1993
Beilstein 1923953
Flash Point 55°C (131°F)
Merck Index 14,9757
Solubility Information Miscible with pentane,ether,toluene and tetrahydrofuran. Sparingly miscible with acetone and acetonitrile. Immiscible with water.
Formula Weight 248.67
Percent Purity 97%
Refractive Index 1.49
Sensitivity Air Sensitive
Density 0.806
Chemical Name or Material Tris(trimethylsilyl)silane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tris(trimethylsilyl)silane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Tris(trimethylsilyl)silane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.