missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyloxy)ethylene, 95%, Thermo Scientific™

Product Code. 10259463 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10259463 1 g Glass bottle 1g
10707022 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10259463 Supplier Thermo Scientific Acros Supplier No. 216010010
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 69097-20-7
Molecular Formula C11H28O3Si3
Molecular Weight (g/mol) 292.60
MDL Number MFCD00011641
InChI Key FCZGHPGTZRTDNN-UHFFFAOYSA-N
Synonym tris trimethylsilyloxy ethylene, tris trimethylsiloxy ethylene, vinyl-2-ylidenetris oxy tris trimethylsilane, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy-3,6-dioxa-2,7-disilaoct-4-ene, 3,6-dioxa-2,7-disilaoct-4-ene, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy, acmc-1b3cp, tris trimethylsiloxy ethene, tris trimethylsiloxy-ethylene, tris-trimethylsiloxy ethylene
PubChem CID 553067
IUPAC Name 1,2-bis(trimethylsilyloxy)ethenoxy-trimethylsilane
SMILES C[Si](C)(C)OC=C(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Colorless to Yellow
Density 0.8850g/mL
Boiling Point 90.0°C (1.0 mmHg)
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)3SiO]2C=CHOSi(CH3)3
Refractive Index 1.4190 to 1.4220
Quantity 1 g
Fieser 08,523; 09,512
Specific Gravity 0.885
Solubility Information Solubility in water: insoluble.
Formula Weight 292.59
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyloxy)ethylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tris(trimethylsilyloxy)ethylene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.