missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Tritest™ pH Paper Roll

Product Code. 10303841 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
1 to 11
Unit Size:
1 roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range unitSize
10303841 1 to 11 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10303841 Supplier Macherey-Nagel™ Supplier No. 90510
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Featuring three indicator zones for the highest accuracy readings. Macherey Nagel™ Tritest™ pH Paper Rolls give the user the most precise readings possible in pH measurement.

  • Provides rapid and reliable results
  • Shows three different colors for each full pH unit
  • Specially equipped with a hydrophobic barrier between the different indicators

Specifications

pH Range 1 to 11
Format Roll
For Use With (Application) For most precise reading these papers show three different colors for each full pH unit
Graduations 1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11
Quantity 1 Each
Type pH Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.