missing translation for 'onlineSavingsMsg'
Learn More

Tritolyl phosphate, mixture of isomers

Product Code. 11448017
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448017 500 mL 500mL
11458017 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448017 Supplier Thermo Scientific Alfa Aesar Supplier No. A17433.AP
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

CAS: 1330-78-5 | C21H21O4P

Tritolyl phosphate, mixture of isomers is used as plasticizer in nitrocellulose, acrylate lacquers, and in polyvinyl chloride. It is a fire retardant for plastics and rubbers. air filter medium, solvent mixtures and heat exchange medium. It is employed as a gasoline additive and as a lead scavenger for tetraethyllead. Further, it acts as a solvent for extraction, especially for nitrocellulose and polymers. In addition to this, it is used for the waterproofing of materials, an antiwear and extreme pressure additive in lubricants and hydraulic fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tritolyl phosphate, mixture of isomers is used as plasticizer in nitrocellulose, acrylate lacquers, and in polyvinyl chloride. It is a fire retardant for plastics and rubbers. air filter medium, solvent mixtures and heat exchange medium. It is employed as a gasoline additive and as a lead scavenger for tetraethyllead. Further, it acts as a solvent for extraction, especially for nitrocellulose and polymers. In addition to this, it is used for the waterproofing of materials, an antiwear and extreme pressure additive in lubricants and hydraulic fluids.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, sulfuric acid, caustics, ammonia, aliphatic amines, alkanolamines, isocyanates, alkylene oxides, epichlorohydrin and nitromethane.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-78-5
Molecular Formula C21H21O4P
MDL Number MFCD02102116
Synonym Tricresyl phosphate; TCP

Specifications

Density 1.167
Boiling Point 265°C (10 mmHg)
Flash Point 250°C (482°F)
Odor Odorless
Linear Formula (CH3C6H4O)3P=O
Quantity 500 mL
UN Number UN2574
Merck Index 14,9763
Solubility Information Miscible with organic solvents. Immiscible with water.
Formula Weight 368.37
Percent Purity 98%
Chemical Name or Material Tritolyl phosphate, mixture of isomers, ortho-isomers <1%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.