missing translation for 'onlineSavingsMsg'
Learn More

Triton™ X-100, Thermo Scientific™

Product Code. 11944741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak Glass Bottle
Glass Bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11944741 10 L Plastic drum 10 L
10591461 250 mL Glass Bottle 250mL
10703411 1 L EcoSafPak Glass Bottle 1 L
10541754 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11944741 Supplier Thermo Scientific Acros Supplier No. 215680100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Polyoxyethylene(10) octylphenyl ether
PubChem CID 5590
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Boiling Point 270.0°C
Melting Point 6.0°C
Density 1.0670g/mL
Quantity 10 L
Flash Point 274°C
Packaging Plastic drum
Refractive Index 1.4900 to 1.4920
Specific Gravity 1.067
Viscosity 240 mPa.s (25°C)
Merck Index 15, 6850
Solubility Information Solubility in water: soluble. Other solubilities: soluble in toluene, xylene, trichloroethylene,, ethylene glycol, ethyl ether, ethyl alcohol,, isopropyl alcohol, ethylene dichloride and most, other solvnets., insoluble in kerosene, miniral spirits.
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
Formula Weight 646.85
Chemical Name or Material Triton™ X-100
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.