missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Triton™ X-405, 70% solution in water

Product Code. 10043623 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10043623 250 mL 250mL
10733601 1 L 1 L
10514664 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10043623 Supplier Thermo Scientific Chemicals Supplier No. 215692500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 9002-93-1 | C16H26O2 | 250.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triton™ X-405
Color Yellow
Name Note 70% Solution in Water
CAS 9002-93-1, 7732-18-5
Physical Form Liquid
Boiling Point 120.0°C
Melting Point -4.0°C
Density 1.0800g/mL
Quantity 250 mL
Flash Point >109°C
Packaging Glass bottle
Refractive Index 1.4360 to 1.4380
Specific Gravity 1.08
Molecular Formula C16H26O2
MDL Number MFCD00132505
Merck Index 15, 6850
Solubility Information Solubility in water: soluble.
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
Molecular Weight (g/mol) 250.38
PubChem CID 5590
Formula Weight 1968.47
Percent Purity ≥65%
Show More Show Less
Product Identifier
  • Triton|TS X-405, 70% solution in water
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.