missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Troglitazone

Product Code. 18065202
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18065202 50 mg 50mg
18045712 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065202 Supplier R&D Systems™ Supplier No. 3114/50
  • ONLINE EXCLUSIVE PRICE
    810.00 €
    765.45 € / 50mg
    Save 44.55 €
    5% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective PPARγ agonist; antidiabetic agent

Selective PPARγ receptor agonist (EC50 values are 780 and 555 nM at murine and human PPARγ receptors respectively). Displays no activity at PPARα or PPARδ receptors. Antidiabetic agent; exerts potent glucose-lowering effects in insulin-resistant diabetic mice. Displays anti-invasive effect on human breast cancer cells; reduces migration, adhesion and spreading on fibronectin-coated plates. Also inhibits cell growth of hematopoietic cell lines. Inhibits lamellipodia formation and actin polymerization.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97322-87-7
Molecular Formula C24H27NO5S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 441.54
Purity 98%
Molecular Weight (g/mol) 441.542
Chemical Name or Material Troglitazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.